Anti-mu Crystallin Rabbit Polyclonal Antibody

Supplier: Proteintech
12495-1-AP-150UL
10090-592EA 402 USD
10090-592
Anti-mu Crystallin Rabbit Polyclonal Antibody
Antibodies
mu Crystallin(thiomorpholine-carboxylate dehydrogenase) is also named as THBP, CRYM, ketimine reductase and belongs to the ornithine cyclodeaminase family. This protein catalyzes the reduction of imine bonds in brain substrates that may include cystathionine ketimine (CysK) and lanthionine ketimine (LK). It is also involved in the regulation of the free intracellular concentration of triiodothyronine and access to its nuclear receptor.

Western Blot:Y79 Cells, 1:1000-1:10000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: mu Crystallin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR