Anti-BMP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
12492-1-AP
10083-296EA 446.09 USD
10083-296
Anti-BMP4 Rabbit Polyclonal Antibody
Antibodies
BMP4, also named as BMP2B and DVR4, belongs to the TGF-beta family. BMPs have been shown to promote astroglial differentiation both in vitro and in vivo. BMP4 have been shown to be components of serum and are thought to be responsible for this differentiation. are expressed mainly in neurons, with minimal expression detected in astrocytes and oligodendrocytes.

Western Blot: Mouse Heart Tissue, 1:200-1:1000; IHC: Human lung Tissue, 1:20-1:200

Type: Primary
Antigen: BMP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR