Anti-COQ6 Rabbit Polyclonal Antibody

Supplier: Proteintech
12481-1-AP-150UL
10085-024EA 402 USD
10085-024
Anti-COQ6 Rabbit Polyclonal Antibody
Antibodies
COQ6(Coenzyme Q10 monooxygenase 6) is also named as CGI-10 and belongs to the ubiH/COQ6 family. It is an evolutionarily conserved flavin-dependent monooxygenase required for biosynthesis of coenzyme Q10, or ubiquinone.It has 3 isoforms produced by alternative splicing and In inner ear, Coq6 is expressed in spiral ganglion, stria vascularis cells, and spiral ligament.Defects in COQ6 are the cause of coenzyme Q10 deficiency, primary, type 6 (COQ10D6).

Western Blot: Mouse Heart Tissue, 1:200-1:1000

Type: Primary
Antigen: COQ6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR