Anti-TPP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12479-1-AP-150UL
10096-232EA 402 USD
10096-232
Anti-TPP1 Rabbit Polyclonal Antibody
Antibodies
TRIPEPTIDYL PEPTIDASE I, TPP1, is a lysosomal exopeptidase that sequentially removes tripeptides from the N termini of proteins. The proenzyme has an apparent molecular mass of 66 kD, and the mature enzyme has an apparent molecular mass of 46 to 48 kD.Defects in TPP1 are the cause of neuronal ceroid lipofuscinosis type 2 (CLN2). This protein distributes widely in all tissues.

Western Blot: Human Colon Tissue, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR