Anti-PREPL Rabbit Polyclonal Antibody

Supplier: Proteintech
12478-1-AP-150UL
10092-774EA 402 USD
10092-774
Anti-PREPL Rabbit Polyclonal Antibody
Antibodies
PREPL(Prolyl endopeptidase-like )is also named as KIAA0436 and belongs to the prolyl oligopeptidase subfamily of serine peptidases.It is probable a serine peptidase whose precise substrate specificity remains unclear. Does not cleave peptides after a arginine or lysine residue.PREPL is a novel oligopeptidase with unique structural and functional characteristics.It has 4 isoforms produced by alternative splicing and can exsit as a homodimer.Defects in PREPL are a cause of hypotonia-cystinuria syndrome (HCS).

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: PREPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR