Anti-LCK Rabbit Polyclonal Antibody

Supplier: Proteintech
12477-1-AP-150UL
10089-556EA 459.7 USD
10089-556
Anti-LCK Rabbit Polyclonal Antibody
Antibodies
LCK(Lymphocyte cell-specific protein-tyrosine kinase) is also named as LSK,p56-LCK and a nonreceptor protein-tyrosine kinase of the SRConcogene family that is involved in transduction of T-cell receptor-mediated activation.It is implicated in the control of lymphocyte growth by virtue of its overexpression in some lymphoid malignancies and its transforming activity in heterologous systems.It has 3 isoforms produced by alternative splicing.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: LCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR