Anti-CDCA8 Rabbit Polyclonal Antibody

Supplier: Proteintech
12465-1-AP-150UL
10084-508EA 402 USD
10084-508
Anti-CDCA8 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Kidney Tissue, 1:200-1:1000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: CDCA8
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR