Anti-VPS34 Rabbit Polyclonal Antibody

Supplier: Proteintech
12452-1-AP-150UL
10081-338EA 402 USD
10081-338
Anti-VPS34 Rabbit Polyclonal Antibody
Antibodies
PIK3C3(Phosphatidylinositol 3-kinase catalytic subunit type 3) is also named as VPS34 and belongs to the PI3/PI4-kinase family.It is a catalytic subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate which plays a key role in initiation and maturation of autophagosomes.It can form a dimer.

Western Blot:PC-3 Cells, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: VPS34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR