Anti-ESAM Rabbit Polyclonal Antibody

Supplier: Proteintech
12445-1-AP
10086-646EA 360.13 USD
10086-646
Anti-ESAM Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human lung Tissue, 1:500-1:5000

Type: Primary
Antigen: ESAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR