Anti-STAM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12434-1-AP-150UL
10095-238EA 453 USD
10095-238
Anti-STAM1 Rabbit Polyclonal Antibody
Antibodies
STAM, also named as STAM1, belongs to the STAM family. It is involved in intracellular signal transduction mediated by cytokines and growth factors. Upon IL-2 and GM-CSL stimulation, it plays a role in signaling leading to DNA synthesis and MYC induction. STAM may also play a role in T-cell development. It is involved in down-regulation of receptor tyrosine kinase via multivesicular body (MVBs). Together with HRS, STAM forms ESCRT-0. STAM was originally identified as an adaptor protein involved in cytokine signaling.

Western Blot:PC-3 Cells, 1:200-1:1000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: STAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR