Anti-HOMER1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12433-1-AP-150UL
10088-394EA 402 USD
10088-394
Anti-HOMER1 Rabbit Polyclonal Antibody
Antibodies
Human HOMER1, also named as SYN47, belongs to the Homer family. Human HOMER1 has some isoforms: HOMER1a, HOMER1b, HOMER1c, HOMER1e and HOMER1h. Mouse Homer1 also named as Vesl-1 and it has some isoforms: Vesl-1l, Vesl-1m, Vesl-1s, Homer1b, and Homer1d. HOMER1 is a postsynaptic density scaffolding protein. It is a neuronal immediate-early gene (IEG) that is enriched at excitatory synapses and binds mGluRs. In WB, the MW of HOMER1 is about 46-50kd, 40-43kd and 20-26kd.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Mouse Brain Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: HOMER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR