Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BBS10 Rabbit Polyclonal Antibody

Supplier: Proteintech

BBS10, also named as C12orf58, belongs to the TCP-1 chaperonin family. It is probable molecular chaperone. Assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex, BBS10 may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. Involved in adipogenic differentiation. Defects in BBS10 are the cause of Bardet-Biedl syndrome type 10 (BBS10).

Western Blot: HepG2 Cells, 1:200-1:2000; IF: HTERT-RPE1 Cells, 1:20-1:200

Type: Primary
Antigen: BBS10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Zebrafish




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 12421-2-AP 10083-518 Each (150µl) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antibody Type Primary
Antigen Name Bardet-Biedl Syndrome 10
Antigen Symbol BBS10
Antigen Synonyms BBS10_HUMAN., FLJ23560, C12orf58, RGD1560748, Bardet Biedl syndrome 10 protein homolog, Bardet Biedl syndrome 10 protein
Clonality Polyclonal
Conjugation Unconjugated
ELISA Yes
Host Rabbit
ImmunoFluorescence Yes
Immunogen Recombinant Protein
Isotype IgG
Purification Antigen affinity Purification
Reactivity Human, Zebrafish
Shipping Temperature Store at -20°C
Storage Buffer PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Western Blot Yes
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty