Anti-PI4KA Rabbit Polyclonal Antibody

Supplier: Proteintech
12411-1-AP-150UL
10092-228EA 402 USD
10092-228
Anti-PI4KA Rabbit Polyclonal Antibody
Antibodies
PI4KA, also named as PIK4, PIK4CA, belongs to the PI3/PI4-kinase family and Type III PI4K subfamily. It acts on phosphatidylinositol (PtdIns) in the first committed step in the production of the second messenger inositol-1,4,5,-trisphosphate. PI4KA plays a role in the formation of membrane complexes where HCV replication takes place. .

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PI4KA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR