Anti-GEMIN4 Rabbit Polyclonal Antibody

Supplier: Proteintech
12408-1-AP-150UL
10087-488EA 402 USD
10087-488
Anti-GEMIN4 Rabbit Polyclonal Antibody
Antibodies
GEMIN4, also named as P97, is a sub-unit of SMN complex. The SMN complex plays a catalyst role in the assembly of small nuclear ribonucleoproteins (snRNPs). It plays an important role in the splicing of cellular pre-mRNAs. The express amount of GEMIN4 is about 8.79 ppm (http://pax-db.org/#!protein/984742). This antibody detects 97-110 kDa GEMIN4.

Western Blot: HEK-293 Cells, 1:200-1:1000; IP: HEK-293 Cells, 1:200-1:1000

Type: Primary
Antigen: GEMIN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR