Anti-MAT1A Rabbit Polyclonal Antibody

Supplier: Proteintech
12395-1-AP-150UL
10089-652EA 402 USD
10089-652
Anti-MAT1A Rabbit Polyclonal Antibody
Antibodies
Methionine adenosyltransferase (MAT) is the enzyme that catalyzes the synthesis of S-adeno-sylmethionine (AdoMet), the main donor of methyl groups in the cell. In mammals MAT is encoded by two different genes, MAT1A and MAT2A. MAT1A is expressed only in the mature liver whereas fetal hepatocytes, extrahepatic tissues and liver cancer cells express MAT2A. This antibody was raised against the full length of human MAT1A and may cross-react with MAT2A. Western blot analysis using this antibody detected a single band around 44-50kD in liver.

Western Blot: Mouse Liver Tissue, 1:1000-1:10000; IHC: Human lung Tissue, 1:20-1:200

Type: Primary
Antigen: MAT1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR