Anti-COPG Rabbit Polyclonal Antibody

Supplier: Proteintech
12393-1-AP-150UL
10084-444EA 402 USD
10084-444
Anti-COPG Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:1000-1:10000; IHC: Human breast cancer, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: COPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR