Anti-UBAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12385-1-AP-150UL
10095-412EA 537.7 USD
10095-412
Anti-UBAP1 Rabbit Polyclonal Antibody
Antibodies
UBAP1, also named as NAG20, is a 55 kDa protein which contains 2 putative tandem UBA domains at the C terminus, a coiled-coil domain, 2 possible N-glycosylation sites, 5 PKC phosphorylation sites, 7 casein kinase II phosphorylation sites, and 3 N-myristoylation sites. UBAP1 might be a potential effective diagnosis candidate for NPC and decreased expression of UBAP1 protein is a possible point of dysfunction along the pathogenesis pathway for NPC that may contribute to malignant transformation.

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human lung cancer, 1:20-1:200; IF: Hela, 1:10-1:100

Type: Primary
Antigen: UBAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR