Anti-UCHL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12384-1-AP-150UL
10096-706EA 599.76 USD
10096-706
Anti-UCHL3 Rabbit Polyclonal Antibody
Antibodies
UCH-L3 belongs to the ubiquitin C-terminal hydrolase family that deubiquitinates ubiquitin-protein conjugates in the ubiquitin-proteasome system. This enzyme indirectly increases the phosphorylation of IGFIR, AKT, FOXO1 and promotes insulin-signaling and insulin-induced adipogenesis. It is required for stress-response retinal, skeletal muscle and germ cell maintenance and may be involved in working memory.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: UCHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR