Anti-PD-ECGF Rabbit Polyclonal Antibody

Supplier: Proteintech
12383-1-AP-150UL
10092-020EA 402 USD
10092-020
Anti-PD-ECGF Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Placenta Tissue, 1:200-1:2000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: PD-ECGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR