Anti-HDGFRP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12380-1-AP-150UL
10087-626EA 459.7 USD
10087-626
Anti-HDGFRP3 Rabbit Polyclonal Antibody
Antibodies
HDGFRP3, also named as Hepatoma-derived growth factor-related protein 3 or HDGF2, is a 203 amino acid protein, which contains 1 PWWP domain and belongs to the HDGF family. HDGFRP3 detected in testis, heart, spinal cord and brain. HDGFRP3 as a nuclear potein enhances DNA synthesis and may play a role in cell proliferation.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200; IP: Mouse Testis Tissue, 1:200-1:1000

Type: Primary
Antigen: HDGFRP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR