Anti-COX2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12375-1-AP-150UL
10093-030EA 459.7 USD
10093-030
Anti-COX2 Rabbit Polyclonal Antibody
Antibodies
COX2 (Prostaglandin G/H synthase 2; PTGS2) mediates the formation of prostaglandins from arachidonate. Its subunit structure is homodimer. The fully N-glycosylated PTGS2 is 72–74 kDa and the aglycosylated is 66 kDa. It also expresses a band of 39 kDa after unspecific cleavage. The 50 kDa band of fragmented PTGS2 has also previously been detected in AD brains.

Western Blot:LPS treated HEK-293 Cells, 1:200-1:2000; IHC: Human ovary Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: COX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR