Anti-IL33 Rabbit Polyclonal Antibody

Supplier: Proteintech
12372-1-AP-150UL
10088-836EA 537.7 USD
10088-836
Anti-IL33 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Transfected HEK-293 Cells, 1:200-1:2000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: IL33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR