PINX1, also named as LPTL, LPTS and Protein 67-11-3, is a Liver-related putative tumor suppressor. It belongs to the PINX1 family. PINX1 is a microtubule-binding protein essential for faithful chromosome segregation. It mediates TRF1 and TERT accumulation in nucleolus and enhances TRF1 binding to telomeres. PINX1 inhibits telomerase activity. It may inhibit cell proliferation and act as tumor suppressor. PINX1 has been identified as a critical telomerase inhibitor and proposed to be a putative tumor suppressor gene. Loss of PinX1 has been found in a large variety of malignancies, but the expression status in epithelial ovarian tumors has not been investigated.
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human skin Tissue, 1:20-1:200
Type: Primary
Antigen: PINX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat