Anti-UQCRH Rabbit Polyclonal Antibody

Supplier: Proteintech
12364-1-AP-150UL
10096-590EA 402 USD
10096-590
Anti-UQCRH Rabbit Polyclonal Antibody
Antibodies
UQCRH(ubiquinol-cytochrome c reductase hinge protein) belongs to the UQCRH/QCR6 family. It is a part of the mitochondrial respiratory chain that may mediate formation of the complex between cytochromes c and c1. The deduced 91-amino acid protein has a 13-amino acid N-terminal mitochondrial targeting presequence.

IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: UQCRH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR