Anti-NDUFB3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12358-1-AP-150UL
10090-966EA 402 USD
10090-966
Anti-NDUFB3 Rabbit Polyclonal Antibody
Antibodies
NDUFB3(NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3) is also named as CI-B12 and belongs to the complex I NDUFB3 subunit family.It is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis.

Western Blot: Human Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: NDUFB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR