Anti-MRRF Rabbit Polyclonal Antibody

Supplier: Proteintech
12357-2-AP-150UL
10090-492EA 402 USD
10090-492
Anti-MRRF Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human lung Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: MRRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR