Anti-TAF12 Rabbit Polyclonal Antibody

Supplier: Proteintech
12353-1-AP-150UL
10095-402EA 402 USD
10095-402
Anti-TAF12 Rabbit Polyclonal Antibody
Antibodies
TAF12, also named TAF15 or TAF2J, is a TBP-associated factor that is contained in Pol I- and Pol II-specific TBP-TAF complexes, it is also a component of the transcription factor IID (TFIID) complex, which is essential for mediating regulation of RNA polymerase transcription. TAF12 plays a key role in regulating eukaryotic gene expression by directly binding promoters and enhancer-bound transactivator proteins. Likewise, TAF12 recruits Gadd45a and the nucleotide excision repair complex to the promoter of rRNA genes leading to active DNA demethylation.

Western Blot:Y79 Cells, 1:200-1:2000

Type: Primary
Antigen: TAF12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR