Anti-S100 A3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12343-1-AP-150UL
10094-084EA 402 USD
10094-084
Anti-S100 A3 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A2780 Cells, 1:200-1:1000

Type: Primary
Antigen: S100 A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR