Anti-PSMD10 Rabbit Polyclonal Antibody

12342-2-AP
10092-964EA 356.38 USD
10092-964
Anti-PSMD10 Rabbit Polyclonal Antibody
Western Blot: Human Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: PSMD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR