Serum deprivation-response protein(SDRP), a substrate of protein kinase C, is a member of a group of proteins that bind to phosphatidylserine in a clacium-dependent manner. SDPR may participate in the generation or stabilization of membrane curvature. SDPR is highly expressed in heart and lung, and low in brain,kidney, liver, pancreas, placenta and skeletal muscle. This is a rabbit polyclonal anti-SDPR antibody raised against C-terminus of human SDPR.
Western Blot:A549 Cells, 1:200-1:2000; IHC: Human Heart Tissue, 1:20-1:200
Type: Primary
Antigen: SDPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat