Anti-ASH2L Rabbit Polyclonal Antibody

Supplier: Proteintech
12331-1-AP-150UL
10082-320EA 402 USD
10082-320
Anti-ASH2L Rabbit Polyclonal Antibody
Antibodies
Western Blot:COS-7 Cells, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: ASH2L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR