Anti-MAP3K7 Rabbit Polyclonal Antibody

Supplier: Proteintech
12330-2-AP-150UL
10095-594EA 524.48 USD
10095-594
Anti-MAP3K7 Rabbit Polyclonal Antibody
Antibodies
MAP3K7(Mitogen-activated protein kinase kinase kinase 7) is also named as TAK1 and belongs to the MAP kinase kinase kinase subfamily.It plays an important role in the cascades of cellular responses evoked by changes in the environment.It has been linked to interleukin-1 receptor and tumor necrosis factor receptor signaling.It has 4 isoforms (53055 kDa; 64-70 kDa and 75-80 kDa)produced by alternative splicing.

Western Blot:CHO Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: MAP3K7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Hamster, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR