Anti-NUP98 Rabbit Polyclonal Antibody

Supplier: Proteintech
12329-1-AP-150UL
10091-482EA 402 USD
10091-482
Anti-NUP98 Rabbit Polyclonal Antibody
Antibodies
Nup98-Nup96 (also known as Nup196, ADIR2 or ADAR2), encoded by NUP98 gene, is a component of the nuclear pore complex (NPC) . It is first synthesized as 186-198 kDa Nup98-Nup96 precursors and the precursors shortly undergo autoproteolysis to give rise to two nucleoporins, the amino-terminal Nup98 (Nup98-N) and carboxy-terminal Nup96 (Nup96-C). Both of Nup98 and Nup96 are localized to the nucleoplasmic side of the NPC. This antibody was raised against the carboxy-terminal region of Nup98-Nup96, it recognizes all Nup98-Nup96 proteins except Nup98-N. Since the autoproteolysis is very rapid this antibody usually detected 98-115 kDa Nup96 protein in multiple cell lines. It is notable that RNA-editing deaminase 1 (Gene ID: 104), also named ADAR2, is a different protein from Nup98-Nup96 (ADAR2).

Western Blot:COLO 320 Cells, 1:500-1:5000

Type: Primary
Antigen: NUP98
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR