Anti-TRIP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
12324-1-AP-150UL
10096-370EA 459.7 USD
10096-370
Anti-TRIP4 Rabbit Polyclonal Antibody
Antibodies
Thyroid hormone receptor interactor 4(TRIP4), other named ASC1, is a transcriptional coactivator that promote transcriptional efficiencies. It harbors an autonomous transactivation domain that contains a putative zinc finger motif, which serves as a binding site for nuclear receptors for thyroid, estrogen, all-trans-retinoic acid and RXR by interaction with SRC-1 or CBP/p300. There are two forms of ASC1 have been detected in mRNA level, isoform1(581aa, ~65kd) and isoform2(539aa,~60kd)(12077347,12390891)

Western Blot: HL-60 Cells, 1:500-1:5000

Type: Primary
Antigen: TRIP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR