Anti-RAB3IP Rabbit Polyclonal Antibody

Supplier: Proteintech
12321-1-AP-150UL
10093-206EA 459.7 USD
10093-206
Anti-RAB3IP Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: Mouse lung Tissue, 1:500-1:5000

Type: Primary
Antigen: RAB3IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR