Anti-NETO2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12318-1-AP
10091-042EA 370.14 USD
10091-042
Anti-NETO2 Rabbit Polyclonal Antibody
Western Blot:SH-SY5Y Cells, 1:200-1:2000

Type: Primary
Antigen: NETO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR