Anti-GNPDA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12312-1-AP-150UL
10087-716EA 459.7 USD
10087-716
Anti-GNPDA1 Rabbit Polyclonal Antibody
Antibodies
GNPDA1(glucosamine-6-phosphate isomerase 1) is also named as GNPI, HLN, KIAA0060 and belongs to the glucosamine/galactosamine-6-phosphate isomerase family. It catalyzes the reversible conversion of D-glucosamine-6-phosphate (GlcN6P) into D-fructose-6-phosphate (Fru6P) and ammonium. It may be playing an essential role for egg activation and early embryo development. GNPI is expressed as a 33-kD cytosolic protein in various cell lines.

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: GNPDA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR