Anti-CHD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12311-1-AP-150UL
10084-692EA 524.48 USD
10084-692
Anti-CHD2 Rabbit Polyclonal Antibody
Antibodies
Chromodomain-helicase-DNA-binding protein 2, also named as ATP-dependent helicase CHD2, CHD2. It is a sequence-selective DNA-binding protein. Molecular mass of CHD2 is 211kD (201kd for isoform 2, 57kd for isoform 3).

Western Blot: Human Placenta Tissue, 1:200-1:2000; IHC: Human Lymphoma, 1:20-1:200

Type: Primary
Antigen: CHD2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR