Anti-GEMIN6 Rabbit Polyclonal Antibody

Supplier: Proteintech
12307-2-AP-150UL
10087-492EA 524.48 USD
10087-492
Anti-GEMIN6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: GEMIN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR