|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ACYP2 |
Antigen Name | Acylphosphatase 2, muscle type |
Reactivity | Mouse, Human, Rat |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | ACYP, ACYM |
Immunogen | Recombinant Protein |
Purification | Antigen affinity Purification |
Storage Buffer | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
Shipping Temperature | Store at -20°C |