Anti-ITGB1BP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12300-1-AP-150UL
10089-018EA 402 USD
10089-018
Anti-ITGB1BP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:PC-3 Cells, 1:200-1:1000; IHC: Human Heart Tissue, 1:20-1:200; IF:PC-3 Cells, 1:10-1:100

Type: Primary
Antigen: ITGB1BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR