|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | ALKBH3 |
Antigen Name | AlkB Homolog 3, Alpha-Ketoglutarate-Dependent Dioxygenase |
Reactivity | Human, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | DEPC-1, ABH3, PCA1, DEPC1 |
Immunogen | Recombinant Protein |
Purification | Antigen affinity Purification |
Storage Buffer | PBS with 0.1% sodium azide and 50% glycerol pH 7.3. |
Shipping Temperature | Store at -20°C |