Anti-CYP4F11 Rabbit Polyclonal Antibody

Supplier: Proteintech
12286-1-AP-150UL
10081-206EA 402 USD
10081-206
Anti-CYP4F11 Rabbit Polyclonal Antibody
Antibodies
CYP4F11, also named as CYPIVF11, belongs to the cytochrome P450 family. It is the lone CYP4F/A enzyme that effectively oxidized 3-hydroxypalmitate. It is the predominant catalyst of this fatty acid oxidation reaction in human liver. CYP4F11 is one of the primary human epidermal keratinocytes. It is positively regulated by multiple signaling pathways in HaCaT keratinocytes, including RXR and JNK signaling pathways.

Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Liver cancer, 1:20-1:200

Type: Primary
Antigen: CYP4F11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR