Anti-PLAC8 Rabbit Polyclonal Antibody

Supplier: Proteintech
12284-1-AP-150UL
10092-424EA 614.88 USD
10092-424
Anti-PLAC8 Rabbit Polyclonal Antibody
Antibodies
PLAC8, also named as Protein C15, is a placenta-specific protein. The expression was downregulated upon activation, particularly in dendritic cells generated in vitro from CD34 (142230) progenitor cells. The MW of PLAC8 is about 12-20kd

IHC: Human tonsil, 1:20-1:200

Type: Primary
Antigen: PLAC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR