Anti-ZWINT Rabbit Polyclonal Antibody

Supplier: Proteintech
12282-2-AP-150UL
10097-408EA 459.7 USD
10097-408
Anti-ZWINT Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human skin Tissue, 1:200-1:1000; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: ZWINT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR