Anti-NDRG4 Rabbit Polyclonal Antibody

Supplier: Proteintech
12184-1-AP-150UL
10090-626EA 459.7 USD
10090-626
Anti-NDRG4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human medulloblastoma, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: NDRG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR