Anti-SMAD5 Rabbit Polyclonal Antibody

Supplier: Proteintech
12167-1-AP-150UL
10094-844EA 459.7 USD
10094-844
Anti-SMAD5 Rabbit Polyclonal Antibody
Antibodies
SMAD5, also named as MADH5 and JV5-1, is a transcriptional modulator activated by BMP (bone morphogenetic proteins) type 1 receptor kinase. SMAD5 is a receptor-regulated SMAD (R-SMAD). It plays a critical role in the signaling pathway by which TGF-beta inhibits the proliferation of human hematopoietic progenitor cells.

Western Blot:K-562 Cells, 1:500-1:5000; IF: MCF-7 Cells, 1:10-1:100; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: SMAD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR