Anti-TCN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12157-1-AP-150UL
10095-736EA 459.7 USD
10095-736
Anti-TCN2 Rabbit Polyclonal Antibody
Antibodies
TCN2 (Transcobalamin-2) is a vitamin B12 (cobalamin; Cbl) binding and transport protein in plasma that facilitates the cellular uptake of the vitamin. TCN2 is secreted in a constitutive fashion in most cells. Plasma TCN2 levels have been reported to be elevated in a number of diverse human diseases. Defects in TCN2 are cause of transcobalamin-2 deficiency. (10714245)

Western Blot:A375 Cells, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TCN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR