Anti-LMBR1L Rabbit Polyclonal Antibody

Supplier: Proteintech
12154-1-AP-150UL
10089-728EA 402 USD
10089-728
Anti-LMBR1L Rabbit Polyclonal Antibody
Antibodies
LMBR1L, also known as LIMR (lipocalin-1-interacting membrane receptor), interacts with lipocalin-1 (Lcn-1), a lipocalin member produced by a number of secretoric glands and tissues and known to bind a variety of lipophilic compounds. LMBR1L is a 489-amino acid protein with a predicted molecular mass of 55 kD and 9 putative transmembrane helices. LMBR1L has been shown to mediate cellular uptake of LMBR1L, thus it is characterized as a endocytic receptor.

Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: LMBR1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR