Anti-PRKCSH Rabbit Polyclonal Antibody

Supplier: Proteintech
12148-1-AP-150UL
10092-782EA 402 USD
10092-782
Anti-PRKCSH Rabbit Polyclonal Antibody
Antibodies
PRKCSH, also named as G19P1, PKCSH and 80K-H, contains a large helix structure and we analyzed mutations in this region using a helical wheel predictor. It is a regulatory subunit of glucosidase II. Defects in PRKCSH are a cause of polycystic liver disease (PCLD).

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: PRKCSH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR